Learn More
Medchemexpress LLC H-His(trt)-OMe hydrochloride | 32946-56-8 | MFCD00237100 | 97.0% | 447.96 g·mol⁻¹ | C26H26ClN3O2 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0222205G
Encompass_Preferred
H-His(trt)-OMe hydrochloride is a protected histidine derivative supplied as the hydrochloride salt and intended for research use. It serves as a building block and intermediate in peptide and organic synthesis, offering selective protection and reactivity for the histidine side chain and a methyl ester C-terminus to facilitate coupling and deprotection strategies.
  • Provides trityl (Trt) protection on the histidine side chain.
  • Contains methyl ester (OMe) at the C-terminus for controlled coupling.
  • Provided as the hydrochloride salt for improved stability and handling.
  • High purity suitable for research and synthesis (97.0%).
  • Useful as an intermediate in peptide synthesis and amino acid derivatization.
Catalog No. 50-003-01347