Learn More
Selleck Chemical LLC H-HoArg-OH
SDP
Supplier:  Selleck Chemical LLC S610925MG
Encompass_Preferred
H-HoArg-OH (L-Homoarginine) is an organ-specific uncompetitive inhibitor of serum bone and liver alkaline phosphatase
Catalog No. 50-000-00788