Learn More
Apexbio Technology LLC H-HoSer-OH 672-15-1 5g
SDP
Supplier:  Apexbio Technology LLC A75835000
Encompass_Preferred
H-HoSer-OH (L-homoserine CAS 672-15-1) is a naturally occurring non-proteinogenic amino acid structurally related to serine distinguished by a -hydroxy side chain It serves as a substrate or intermediate in various enzymatic processes contributing to amino acid metabolism and biosynthetic pathways Research indicates that homoserine participates in neurotransmitter metabolic routes thereby modulating neural function Owing to its distinct side chain H-HoSer-OH is frequently utilized in the synthesis of peptides and their derivatives for probing the biological significance of amino acid modifications within proteins
Catalog No. 50-000-93166