Learn More
STA PHARMACEUTICAL US LLC H-L-Lys(Fmoc)-OMe HCl | 25 g | CAS 201009-98-5 | MDL MFCD00153447
Supplier: STA PHARMACEUTICAL US LLC STAAA1082225GM
H-L-Lys(Fmoc)-OMe HCl is a Amino Acid reagent (Subcategory: Lys) sold by WuXi TIDES. Offered in 25 g. Store at 4 °C. SDS available for reference.
Specifications
- CAS: 201009-98-5
- MDL: MFCD00153447
- InChIKey: KKSJCURWQOUTKB-BDQAORGHSA-N
- Molecular Weight: 418.918
- Molecular Formula: C22H27ClN2O4
- Purity: ≥95%
- Container Type: 125 mL HDPE
- Pack Size: 25 g
- Net Weight: 25 g
- Gross Weight: 54.3 g
- Commodity Code: 29242970
- Country Of Origin: China
- IUPAC: methyl N6-(((9H-fluoren-9-yl)methoxy)carbonyl)-L-lysinate hydrochloride
- SMILES: COC([C@H](CCCCNC(OCC1C2=CC=CC=C2C3=CC=CC=C31)=O)N)=O.Cl
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.