Learn More
Medchemexpress LLC H-Lys-Trp-Lys-OH | 38579-27-0 | 98.3% | 460.57 | 50 MG

Supplier: Medchemexpress LLC HYP135050MG
H-Lys-Trp-Lys-OH is a small molecule peptide, identified as Compound AMP12 and extracted from patent CN 104072579 A. This compound demonstrates significant antibacterial and antiviral activities, making it suitable for various research applications.
- Displays antibacterial activity
- Displays antiviral activity
- Molecular weight: 460.57
- Formula: C23H36N6O4
- CAS number: 38579-27-0
- Appears as a white to off-white solid
- Soluble in H2O at 100 mg/mL, requiring sonication
- Recommended solid storage: -80°C for 2 years, or -20°C for 1 year
- Recommended storage in solvent: -80°C for 6 months, or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.