Learn More
Medchemexpress LLC H-Lys(Z)-OH | 1155-64-2 | 280.32 | 100 G

Supplier: Medchemexpress LLC HYW007750100G
H-Lys(Z)-OH is a lysine derivative. Amino acids and their derivatives, like H-Lys(Z)-OH, are recognized for their benefits as ergogenic dietary substances. They play a role in influencing the secretion of anabolic hormones, providing fuel during exercise, enhancing mental performance under stress, and preventing exercise-induced muscle damage.
- Compound type: Lysine derivative
- For research purposes only
- Influences anabolic hormone secretion
- Provides fuel during exercise
- Aids mental performance
- Prevents muscle damage
- Appearance: Solid, white to off-white
- Powder stored at -20°C for 3 years or 4°C for 2 years
- In solvent stored at -80°C for 6 months or -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.