Learn More
BIOHIPPO h/m FOXA1-sh-GFP-Puro
SDP
Supplier:  BIOHIPPO BHV194001723
Encompass
h/m FOXA1-sh-GFP-Puro shRNA lentiviral vector expressing GFP with Puromycin selection for studies in Human/Mouse Each vial provides a nominal titer of 5x10E6 transducing units TU Validated h/m FOXA1 shRNA Lentivirus High-titer shRNA lentiviral particles specific for human/mouse forkhead box A1 (FOXA1) The shRNA has been validated to meet or exceed 70% knockdown of FOXA1 using a specific fluorescence-based method that is more rapid and reliable than qPCR The lentiviral particles are ultra-purified and concentrated by PEG precipitation and sucrose gradient centrifugation and are ideal for transducing difficult-to-transfect cells including thawed and/or primary cells Order a shRNA lentivirus set and receive lentivirus produced from mix of 2 independent shRNAs validated to knockdown the target gene ( 70%) plus control lentivirus produced from mix of 2 scrambled shRNA constructs
Catalog No. 50-003-31001