Learn More
Bachem H-Met-AMC
SDP
Supplier:  Bachem 4002590.0250
Encompass_Preferred
H-Met-AMC Acetate, 94367-34-7, C15H18N2O3S, Mr 306.39, 250mg. Fluorogenic substrate for methionine aminopeptidase 2, Km 0.31 ± 0.02 mM and kcat 3.3 ± 0.1 min-1 at pH 7.5 and 30°C.
Catalog No. 50-260-295