Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC H-Phe(3,5-DiF)-OH | 31105-91-6 | 201.17 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0060691G

Encompass_Preferred

H-Phe(3,5-DiF)-OH is a difluorophenylalanine in the L-configuration [L-(F2)Phe]. It can be incorporated into the thrombin receptor-tethered ligand peptide SFLLRNP to identify the phenyl hydrogens of the Phe-2 residue involved in the CH/π receptor interaction. This product appears as a white to off-white solid with a molecular weight of 201.17.

  • Difluorophenylalanine in the L-configuration
  • Can be incorporated into thrombin receptor-tethered ligand peptide SFLLRNP
  • Used to identify phenyl hydrogens of Phe-2 residue involved in CH/π receptor interaction
  • For research use only
  • Available documentation: data sheet, SDS, COA, HNMR, RP-HPLC, NP-HPLC, handling instructions
  • Powder storage: -20°C for 3 years; 4°C for 2 years
  • In solvent storage: -80°C for 6 months; -20°C for 1 month

Catalog No. 50-004-47355


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.