Learn More
Medchemexpress LLC H-Phe(3-Cl)-OH | 80126-51-8 | 98.59% | 199.64 | 25 G

Supplier: Medchemexpress LLC 25GHYW012487
H-Phe(3-Cl)-OH is a phenylalanine derivative. Amino acids and amino acid derivatives, such as H-Phe(3-Cl)-OH, are commercially used as ergogenic supplements that influence the secretion of anabolic hormones, supply of fuel during exercise, mental performance during stress-related tasks, and prevent exercise-induced muscle damage. These substances are recognized as beneficial ergogenic dietary substances.
- For research use only.
- Target: amino acid derivatives.
- Pathway: others.
- Store powder at -20°C for 3 years or 4°C for 2 years.
- Store in solvent at -80°C for 6 months or -20°C for 1 month.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.