Learn More
Medchemexpress LLC H-Phe-Asp-OH | 22828-05-3 | 99.1% | 280.28 | 25 MG

Supplier: Medchemexpress LLC 25MGHYP4472
H-Phe-Asp-OH is a polypeptide identified through peptide screening, a research tool that uses immunoassays to pool active peptides. It is utilized for protein interaction, functional analysis, and epitope screening, particularly in agent research and development.
- Polypeptide
- High purity (99.08%)
- Research tool for peptide screening, protein interaction, functional analysis, and epitope screening
- Multiple package sizes
- Solid appearance
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.