Learn More
Medchemexpress LLC H-Phe-Phe-OH | 2577-40-4 | 312.37 | 1 G

Supplier: Medchemexpress LLC HYW0079701G
H-Phe-Phe-OH is a peptide composed of two phenylalanine molecules. Phenylalanine is an essential amino acid and a precursor for the amino acid tyrosine and catecholamines. The H-Phe-Phe-OH recognition motif of the Alzheimer's Abeta peptide is the smallest peptide capable of assembling into higher-order structures.
- Purity: 99.7%
- Molecular weight: 312.37
- CAS number: 2577-40-4
- Appears as a white to off-white solid
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.