Learn More
Medchemexpress LLC H-Phe-Phe-OH | 2577-40-4 | 312.37 | 50 MG

Supplier: Medchemexpress LLC HYW00797050MG
H-Phe-Phe-OH is a peptide composed of two phenylalanine molecules, with phenylalanine serving as an essential amino acid and a precursor for tyrosine and catecholamines. It is also a constituent in psychotropic drugs and its recognition motif is significant in the study of Alzheimer's Abeta peptide assembly.
- Chemical formula: C18H20N2O3
- Appearance: Solid
- Color: White to off-white
- Derived from endogenous metabolite
- Store powder at -20°C for 3 years or 4°C for 2 years
- Store in solvent at -80°C for 6 months or -20°C for 1 month
- Soluble in 80% Acetic acid/water (50 mg/mL)
- Slightly soluble in DMSO
- Insoluble in water
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.