Learn More
Medchemexpress LLC H-Phe-Phe-OH | 2577-40-4 | MFCD00063154 | 312.37 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0079705G
Encompass_Preferred
H-Phe-Phe-OH is a peptide composed of two phenylalanine molecules. Phenylalanine is an essential amino acid, serving as a precursor for tyrosine and catecholamines in the body. It is also a constituent in various psychotropic drugs.
  • Composed of two phenylalanine molecules
  • Precursor for tyrosine
  • Precursor of catecholamines in the body
  • Constituent in psychotropic drugs
  • Recognition motif of the Alzheimer's Abeta peptide
  • Smallest peptide known to assemble into higher-order structures
  • For research use only
Catalog No. 50-004-46613