Learn More
Selleck Chemical LLC H-Phe-Phe-OH
SDP
Supplier:  Selleck Chemical LLC S609350MG
Encompass_Preferred
H-Phe-Phe-OH (Phenylalanyl-phenylalanine Phe-Phe L-Phe-L-Phe) is a short peptide diphenylalanine with utility for potential applications in biomaterial chemistry sensors and bioelectronics
Catalog No. 50-000-03018