Learn More
Bachem H-Phe-Phe-OH
SDP
Supplier:  Bachem 4000605.0005
Encompass_Preferred
H-Phe-Phe-OH, 2577-40-4, C18H20N2O3, Mr 312.37, 5g. When evaporating HFIP solutions of diphenylalanine, nanotubes are formed. These structures have been used for casting silver nanowires, the dipeptide is removed from the wires by enzymatic degradation.
Catalog No. 50-259-373