Learn More
Apexbio Technology LLC H-Phe-pNA 2360-97-6 1g
SDP
Supplier:  Apexbio Technology LLC A72701000
Encompass_Preferred
H-Phe-pNA (nan) is a synthetic chromogenic substrate commonly utilized for assessing the catalytic activity of serine proteases particularly chymotrypsin and structurally related enzymes It is used to monitor enzyme kinetics and proteolytic reactions by releasing the chromophore para-nitroaniline (pNA) upon enzymatic cleavage H-Phe-pNA exerts its utility as a biochemical tool by permitting real-time measurement of protease activity via spectrophotometric detection at 405 nm Based on these properties H-Phe-pNA holds research potential in enzyme activity quantification functional characterization of proteases and high-throughput screening of potential inhibitors in drug discovery and biomedical research
Catalog No. 50-224-0498