Learn More
Medchemexpress LLC H-Phg(4-Cl)-OH (L-4-chlorophenylglycine) | 67336-19-0 | 185.61 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW00572010G
Encompass_Preferred
H-Phg(4-Cl)-OH (L-4-chlorophenylglycine) is a glycine derivative recognized as a beneficial ergogenic dietary substance. As an amino acid derivative, it can influence the secretion of anabolic hormones, supply fuel during exercise, improve mental performance during stress-related tasks, and help prevent exercise-induced muscle damage.
  • Glycine derivative
  • Functions as an ergogenic supplement
  • Influences anabolic hormone secretion
  • Supplies fuel during exercise
  • Improves mental performance during stress-related tasks
  • Helps prevent exercise-induced muscle damage
  • White to off-white solid appearance
  • High purity (98.9%)
  • Molecular weight of 185.61
Catalog No. 50-004-49311