Learn More
Medchemexpress LLC H-Phg(4-Cl)-OH (L-4-Chlorophenylglycine) | 67336-19-0 | 185.61 | 25 G

Supplier: Medchemexpress LLC 25GHYW005720
H-Phg(4-Cl)-OH (L-4-Chlorophenylglycine) is a Glycine derivative and an amino acid derivative. Amino acids and their derivatives are commercially used as ergogenic supplements, influencing anabolic hormone secretion, fuel supply during exercise, mental performance during stress, and preventing exercise-induced muscle damage. They are recognized as beneficial ergogenic dietary substances.
- Molecular formula: C8H8ClNO2
- Appearance: White to off-white solid
- Powder storage: -20°C for 3 years, 4°C for 2 years
- In solvent storage: -80°C for 6 months, -20°C for 1 month
- 1H NMR spectrum consistent with structure
- Tested and complies with specifications
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.