Learn More
Medchemexpress LLC H-Pro-Gly-Pro-OH | 7561-51-5 | 963960 | 99.79% | 269.30 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP44865MG
Encompass_Preferred
H-Pro-Gly-Pro-OH is a collagen-derived matrikine that acts as a neutrophil chemoattractant, directing a localized repair response. It also activates the transcription of neurotrophins and their receptor genes after cerebral ischemia.
  • Collagen-derived matrikine
  • Neutrophil chemoattractant
  • Activates neurotrophin transcription
  • Involved in localized repair response
  • Suitable for research applications
Catalog No. 50-003-97676