Learn More
Bachem H-Pro-Ile-OH
SDP
Supplier:  Bachem 4001060.0001
Encompass_Preferred
H-Pro-Ile-OH, 51926-51-3, C11H20N2O3, Mr 228.29, 1g. Specific inhibitor of rat endopeptidase 24.16 (oligopeptidase M, neurolysin).
Catalog No. 50-259-376