Learn More
Apexbio Technology LLC H-Pro-OtBu 2812-46-6 5g
SDP
Supplier:  Apexbio Technology LLC A72815000
Encompass_Preferred
H-Pro-OtBu is an amino acid derivative frequently employed in peptide synthesis structural biology studies and pharmaceutical research Structurally the compound consists of a proline residue protected by a tert-butyl ester functional group In both solid-phase and solution-phase peptide synthetic strategies H-Pro-OtBu serves as an intermediate that facilitates the selective introduction of proline residues into peptide chains Additionally researchers utilize this derivative in studies investigating peptide and protein conformations particularly regarding proline-rich sequences or cyclic peptides to elucidate structure-function correlations and inform therapeutic compound design
Catalog No. 50-224-0852