Learn More
Medchemexpress LLC H-Pro-Val-OH | 52899-09-9 | 99.3% | 214.26 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP4566
Encompass_Preferred
H-Pro-Val-OH is a deprotonation dipeptide that contains proline. It can catalyze the Michael addition reaction of acetone to trans-β-nitrostyrene. This dipeptide also serves as a substrate for fibroblast enzymes and prolinase, offering potential applications in various biochemical analyses.
  • Contains proline
  • Catalyzes Michael addition reaction of acetone to trans-β-nitrostyrene
  • Serves as a substrate for fibroblast enzymes and prolinase
  • Useful in biochemical analysis
Catalog No. 50-003-97235