Learn More
Genetex Inc Helicobacter pylori Catalase Monoclonal Antibody [MSVA-466M] HistoMAX™ (GTX04479) GeneTex - IHC-Optimized Antibody.
SDP
Supplier:  Genetex Inc GTX04479
Encompass_Preferred
The Helicobacter pylori Catalase antibody [MSVA-466M] HistoMAX™ (GTX04479) is a Mouse Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human stomach tissue using GTX04479 Helicobacter pylori Catalase antibody [MSVA-466M] HistoMAX. Complete absence of helicobacter pylori in a stomach biopsy.
Catalog No. NC3667789