Learn More
Medchemexpress LLC H-Ser-His-OH | 67726-09-4 | 99.8% | 10 MG

Supplier: Medchemexpress LLC HY12648810MG
H-Ser-His-OH is a short peptide identified as an endogenous metabolite with hydrolysis cleavage activity.
- Short peptide
- Hydrolysis cleavage activity
- Endogenous metabolite
- Molecular weight of 242.23
- Chemical formula: C9H14N4O4
- Appears as a white to off-white solid
- Soluble in water and DMSO
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.