Learn More
Carbosynth LLC. H-Ser-Leu-Ile-Gly-Arg-Leu-NH2, 1mg, MW: 656.83, Molecular formula: C29H56N10O7, CAS# 171436-38-7
SDP
Supplier:  Carbosynth LLC. PAR3664PI1MG
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements
Synonyms: H-Ser-Leu-Ile-Gly-Arg-Leu-NH2, SLIGRL-Amide, Application: Protease-Activated Receptor 2 (PAR2) Agonist, Salt Form:, Acetate Salt, Storage Temperature: -20C, References: B. Al-Ani, M. Saifeddine, and M.D. Hollenberg, Can. J. of Physiol. Pharmacol., 73, 1203 (1995)., N. Vergnolle, Br. J. Pharmacol., 141, 1264 (2004). (Review)
Catalog No. 50-168-6174