Learn More
Carbosynth LLC. H-Ser-Phe-Leu-Leu-Arg-Asn-NH2, 5mg, MW: 747.9, Molecular formula: C34H57N11O8, CAS# 141923-40-2
SDP
Supplier:  Carbosynth LLC. PAR3676PI5MG
Encompass_Preferred
Synonyms: H-Ser-Phe-Leu-Leu-Arg-Asn-NH2, SFLLRN-Amide, Application: Selective Protease-Activated Receptor 1 (PAR1) Agonist, Salt Form:, Acetate Salt, Storage Temperature: -20°C, References: R.R. Vassallo, Jr., T. Kieber-Emmons, K. Cichowski, and L.F. Brass, J. Biol. Chem., 267, 6081 (1992)., N. Vergnolle, Br. J. Pharmacol., 141, 1264 (2004). (Review)
Catalog No. 50-168-6187