Learn More
Medchemexpress LLC H-Ser-Phe-OH | 16875-28-8 | 99.8% | 252.27 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP4509100MG
Encompass_Preferred
H-Ser-Phe-OH is a polypeptide identified through peptide screening, a research tool that uses immunoassay to pool active peptides. It is utilized for protein interaction, functional analysis, and epitope screening, particularly in agent research and development.
  • Polypeptide
  • Used in peptide screening
  • Supports protein interaction studies
  • Enables functional analysis
  • Suitable for epitope screening
Catalog No. 50-003-98750