Learn More
Medchemexpress LLC H-Thr(tBu)-OMe HCl | 71989-43-0 | MFCD00077111 | 98.0% | 225.71 g/mol | C9H20ClNO3 | 100 G

Supplier: Medchemexpress LLC HYW042013100G
H-Thr(tBu)-OMe HCl is a protected threonine methyl ester hydrochloride used as an amino acid derivative building block in peptide synthesis and organic chemistry. It is supplied as the hydrochloride salt and is commonly used when side-chain protection of threonine is required.
- Cas number: 71989-43-0.
- Molecular weight 225.71 g/mol.
- Chemical formula C9H20ClNO3.
- Purity typically 98.0%.
- Used as a building block in peptide synthesis and organic synthesis.
- Available in laboratory quantities including 100 g.
- Provided as the hydrochloride salt for stability and ease of handling.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.