Learn More
Medchemexpress LLC H-Thr(tBu)-OMe hydrochloride | 71989-43-0 | MFCD00077111 | ≥98.0% | 225.71 g·mol⁻¹ | C9H20ClNO3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW04201310G
Encompass_Preferred
H-Thr(tBu)-OMe hydrochloride is the tert-butyl protected methyl ester of threonine supplied as its hydrochloride salt. It is used as an amino acid building block in peptide synthesis and in synthetic organic chemistry for incorporation of protected threonine residues. The compound has CAS 71989-43-0, a molecular weight of 225.71 g·mol⁻¹, and is typically supplied with a purity of ≥98.0%.
  • Protected methyl ester of threonine.
  • Hydrochloride salt improves stability and handling.
  • Suitable for peptide synthesis and small-molecule synthesis.
  • High purity (≥98.0%) as supplied.
  • Available in multiple pack sizes, including 10 g.
Catalog No. 50-002-09252