Learn More
Cayman Chemical H TOPK 032 1mg
SDP
Supplier:  Cayman Chemical 198151 MG
Encompass_Preferred
An inhibitor of TOPK (IC50 = ~2 µM), Chk1 (IC50 = 9.6 µM), and MEK1 (40% inhibition at 5 µM); decreases the growth of colon cancer and glioma initiating cells in vitro and suppresses tumor growth in vivo
Catalog No. 50-298-1554