Learn More
Medchemexpress LLC H-Trp-Glu-OH | 36099-95-3 | 99.95% | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY128487
Encompass_Preferred
H-Trp-Glu-OH is a selective, reversible, and cell-permeable PPARγ antagonist. It has a Kd of approximately 8 μM and may be developed as a potential lead compound in diabetes research.
  • Selective, reversible, and cell-permeable PPARγ antagonist.
  • Kd of approximately 8 μM.
  • Potential lead compound in diabetes research.
Catalog No. 50-003-42412