Learn More
Medchemexpress LLC H-Trp-NH2.HCl | 5022-65-1 | 99.95% | 239.70 | 10 G

Supplier: Medchemexpress LLC HYW00876610G
H-Trp-NH2.HCl is an endogenous metabolite primarily used as a laboratory chemical for research purposes. It is suitable for the manufacture of substances and intended for experienced personnel only, not for medical applications.
- Endogenous metabolite
- Off-white to gray solid appearance
- Soluble in DMSO and H2O
- Store at 4°C, sealed, away from moisture
- Store in solvent at -80°C for 6 months; -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.