Learn More
Medchemexpress LLC H-Trp-Phe-OH | 6686-02-8 | 99.87% | 351.40 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP464110MG
Encompass_Preferred
H-Trp-Phe-OH is a dipeptide composed of tryptophan and phenylalanine (Trp-Phe). It acts as an antihypertensive peptide by inhibiting angiotensin-converting enzyme (ACE), leading to increased nitric oxide (NO) levels and decreased endothelin-1 (ET-1) levels. Studies have also indicated its ability to increase ovarian weight in female mice.
  • Antihypertensive peptide
  • Inhibits angiotensin-converting enzyme (ACE)
  • Increases nitric oxide (NO) levels
  • Decreases endothelin-1 (ET-1) levels
  • May increase ovarian weight
Catalog No. 50-003-98558