Learn More
Medchemexpress LLC H-Trp-Phe-OH | 6686-02-8 | 99.9% | 351.40 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP4641100MG
Encompass_Preferred
H-Trp-Phe-OH is a dipeptide composed of tryptophan and phenylalanine (Trp-Phe). It functions as an antihypertensive peptide by inhibiting angiotensin-converting enzyme (ACE), and it also increases NO levels and decreases endothelin-1 (ET-1) levels in a dose-dependent manner. This peptide has also been observed to increase ovarian weight in female mice.
  • Dipeptide composed of tryptophan and phenylalanine
  • Functions as an antihypertensive peptide
  • Inhibits angiotensin-converting enzyme (ACE)
  • Increases NO levels dose-dependently
  • Decreases endothelin-1 (ET-1) levels
Catalog No. 50-004-00059