Learn More
Medchemexpress LLC H-Trp-Trp-Trp-OH | 59005-82-2 | 99.6% | 576.64 | 10 MG

Supplier: Medchemexpress LLC HYP464810MG
H-Trp-Trp-Trp-OH is a tripeptide composed of tryptophan that exhibits antibacterial activity.
- Tripeptide consisting of tryptophan.
- Has antibacterial activity.
- Appears as a solid.
- Color is white to off-white.
- Molecular weight is 576.64.
- Formula is C33H32N6O4.
- Used in infection and bacterial infection research.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.