Learn More
Medchemexpress LLC H-Trp-Trp-Trp-OH | 59005-82-2 | >99.6% | 576.64 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYP4648100MG
Encompass_Preferred
H-Trp-Trp-Trp-OH is a tripeptide composed of tryptophan and exhibits antibacterial activity. This product is intended for research and analytical applications.
  • Tripeptide composed of tryptophan
  • Exhibits antibacterial activity
  • Powder can be stored at -80°C for 2 years or -20°C for 1 year
  • In solvent, can be stored at -80°C for 6 months or -20°C for 1 month
  • Requires sealed storage, away from moisture
Catalog No. 50-003-99224