Learn More
Medchemexpress LLC H-Tyr(3-I)-OH | 70-78-0 | 307.09 | 10 G

Supplier: Medchemexpress LLC HYW00845210G
H-Tyr(3-I)-OH is a potent and effective tyrosine hydroxylase inhibitor. It is an intermediate in the production of thyroid hormones and acts as a human or mouse metabolite.
- Potent and effective tyrosine hydroxylase inhibitor
- Intermediate in thyroid hormone production
- Functions as a human or mouse metabolite
- High purity of 99.93%
- White to off-white solid appearance
- Inhibits enzymatic activity by 100% at 100 μM
- Inhibits enzymatic activity by 60-70% at 10 μM
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.