Learn More
Medchemexpress LLC H-Tyr-Ile-Gly-Ser-Arg-NH2 | 110590-65-3 | 98.3% | 593.68 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP46511MG
Encompass_Preferred
H-Tyr-Ile-Gly-Ser-Arg-NH2 is a hexapeptide that is an essential fragment of laminin, responsible for efficient cell adhesion and receptor binding. It promotes various epithelial cell adhesion activities.
  • Essential fragment of laminin
  • Responsible for efficient cell adhesion
  • Responsible for receptor binding
  • Promotes various epithelial cell adhesion activities
Catalog No. 50-003-96699