Learn More
Medchemexpress LLC H-Tyr-Ile-Gly-Ser-Arg-NH2 | 110590-65-3 | 98.3% | 593.68 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP465110MG
Encompass_Preferred
H-Tyr-Ile-Gly-Ser-Arg-NH2 is a hexapeptide that is an essential fragment of laminin responsible for efficient cell adhesion and receptor binding. It promotes various epithelial cell adhesion activities.
  • Essential fragment of laminin
  • Mediates cell adhesion
  • Binds receptors
  • Promotes epithelial cell adhesion activities
  • Targets TGF-β receptor
  • Involved in TGF-beta/Smad pathway
Catalog No. 50-003-96570