Learn More
Medchemexpress LLC H-Tyr-Tyr-OH 10mM 1mL | 1050-28-8 | 344.36 g/mol | C18H20N2O5 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY11478210MM 1ML
Encompass_Preferred
H-Tyr-Tyr-OH (L-tyrosyl-L-tyrosine) is a dipeptide research reagent reported to inhibit angiotensin I-converting enzyme (ACE). It is supplied as a 10 mM solution in DMSO (1 mL) and is also available as solid quantities; reported purity is 99.02% and molecular weight is 344.36 g/mol.
  • ACE inhibitor activity useful for blood pressure and enzymatic studies.
  • Available as a ready-to-use 10 mM solution in DMSO for quick assays.
  • Solid form available for custom formulation and longer-term storage.
  • High reported purity (99.02%) suitable for analytical and biological experiments.
  • Molecular weight (344.36 g/mol) provided for accurate dosing and calculations.
Catalog No. 50-002-29644