Learn More
Medchemexpress LLC H-Val-Ala-OH (Standard) | 27493-61-4 | 188.23 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYW00703510MM/1ML
Encompass_Preferred
H-Val-Ala-OH (Valyl-alanine) is a dipeptide formed from L-Valine and L-Alanine residues that functions as a metabolite and possesses a bitter taste. This compound exhibits the ability to self-assemble into various ordered structures, such as square plates and wires, depending on the solvent, concentration, and intermolecular hydrogen bonds.
  • Dipeptide formed from L-Valine and L-Alanine residues
  • Functions as a metabolite
  • Possesses a bitter taste
  • Self-assembles into ordered structures
  • Used for research applications only
Catalog No. 50-004-60276