Learn More
Medchemexpress LLC H-Val-Ala-OH (Valyl-alanine) | 27493-61-4 | 188.23 | 100 G

Supplier: Medchemexpress LLC HYW007035100G
H-Val-Ala-OH (Valyl-alanine) is a dipeptide formed from L-Valine and L-Alanine residues. It functions as a metabolite and is noted for its bitter taste.
- Molecular weight: 188.23
- Formula: C8H16N2O3
- Appearance: Solid
- Color: White to off-white
- Smiles: C[C@@H](C(O)=O)NC([C@H](C(C)C)N)=O
- Structure classification: Ketones, Aldehydes, Acids
- Initial source: Endogenous metabolite
- Solubility (In Vitro): H2O: 100 mg/mL (531.26 mM; Requires ultrasonic), DMSO: < 1 mg/mL (insoluble or slightly soluble)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.