Learn More
Apexbio Technology LLC H-Val-OtBu.HCl 13518-40-6 25g
SDP
Supplier:  Apexbio Technology LLC A744125000
Encompass_Preferred
H-Val-OtBu HCl (CAS 13518-40-6) is a tert-butyl ester hydrochloride derivative of the amino acid valine It is commonly employed as an intermediate reagent in peptide synthesis and medicinal chemistry research The compound s protected carboxyl group allows controlled and selective coupling reactions at the amino terminus facilitating the elongation of peptide chains Based on these properties H-Val-OtBu HCl holds research potential in the synthesis of linear and cyclic peptides as well as in structure-activity relationship investigations and rational development of peptide-based pharmaceutical precursor molecules
Catalog No. 50-000-94059