Learn More
Medchemexpress LLC H-Val-Pro-Pro-OH | 58872-39-2 | 99.8% | 311.38 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY11416110MG
Encompass_Preferred
H-Val-Pro-Pro-OH is a milk-derived proline peptides derivative and an inhibitor of Angiotensin I converting enzyme (ACE), with an IC50 of 9 μM. It enhances insulin sensitivity and prevents insulin resistance in 3T3-F442A pre-adipocytes, and also has anti-hypertensive and anti-inflammatory functions. It further enhances the expression of glucose transporter 4 (GLUT4) in adipocytes and restores glucose uptake in TNF-treated adipocytes.
  • Inhibits Angiotensin I converting enzyme (ACE) with an IC50 of 9 μM.
  • Enhances insulin sensitivity.
  • Prevents insulin resistance in 3T3-F442A pre-adipocytes.
  • Has anti-hypertensive functions.
  • Has anti-inflammatory functions.
  • Enhances the expression of glucose transporter 4 (GLUT4) in adipocytes.
  • Restores glucose uptake in TNF-treated adipocytes.
Catalog No. 50-004-35647