Learn More
Medchemexpress LLC H-Val-Pro-Pro-OH | 58872-39-2 | MFCD08458654 | 99.9% | 311.38 | C15H25N3O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1141615MG
Encompass_Preferred
H-Val-Pro-Pro-OH is a milk-derived tripeptide that inhibits angiotensin I-converting enzyme (ACE) with a reported IC50 of approximately 9 μM. It has been reported to exhibit anti-hypertensive and anti-inflammatory effects, and to enhance GLUT4 expression and glucose uptake in adipocyte models. The compound is supplied as a white to off-white solid with high purity and recommended frozen storage for long-term stability.
  • Inhibits angiotensin I-converting enzyme (ACE) with IC50 ≈ 9 μM.
  • Reported anti-hypertensive and anti-inflammatory activity.
  • Enhances GLUT4 expression and restores glucose uptake in adipocytes.
  • High purity solid suitable for biochemical and cellular assays.
  • Soluble in DMSO (12.5 mg/mL) with sonication and warming to 60°C.
  • Stable under recommended frozen storage conditions for long-term use.
Catalog No. 50-002-63878