Learn More
Bachem (Arg13)-Amyloid Beta Protein (1-40) Trifluoroacetate
SDP
Supplier:  Bachem 4095736.0500
Encompass_Preferred
(Arg13)-Amyloid Beta Protein (1-40) Trifluoroacetate, H-Asp-Ala-Glu-Phe-Arg-His-Asp-Ser-Gly-Tyr-Glu-Val-Arg-His-Gln-Lys-Leu-Val-Phe-Phe-Ala-Glu-Asp-Val-Gly-Ser-Asn-Lys-Gly-Ala-Ile-Ile-Gly-Leu-Met-Val-Gly-Gly-Val-Val-OH, 1816939-33-9, C194H300N54O58S, Mr 4348.91, 0.5mg. [Arg13] b-Amyloid (1-40). H13R, a mutation in the metal-binding region of Abeta reduces its copper-mediated toxicity. The native rodent sequence containing an arginine at this position is more tolerant to metals than the human amyloid peptide.
Catalog No. 50-194-6575