Learn More
Sino Biological Influenza A H1N1 (A/Wisconsin/67/2022) Hemagglutinin / HA Trimer Protein (His Tag) (MALS-verified)
Supplier:  Sino Biological 40940V08H7100
Encompass_Preferred
A DNA sequence encoding the Influenza A virus ((A/Wisconsin/67/2022) (H1N1)) Hemagglutinin / HA (translated amino acids of EPI2224788), termed as HA, was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3549