Learn More
Sino Biological Influenza A H1N1 (A/Victoria/2570/2019) Hemagglutinin / HA-specific B cell probe Protein (Y108F, AVI & His Tag), Biotinylated (HPLC-verified)
Supplier:  Sino Biological 41050V27HB20
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/Victoria/2570/2019 (H1N1)) hemagglutinin (EPI2413494) (1-519, with a mutation Tyr108Phe),was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-299-3369