Learn More
Medchemexpress LLC H2-003 | 1060438-30-3 | >99.5% | 446.50 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY231991MG
Encompass_Preferred
H2-003 is a selective human DGAT2 inhibitor that effectively inhibits triglyceride (TG) biosynthesis and lipid droplet formation in 3T3-L1 cells. It is suitable for research on DGAT2 and TG-related metabolic diseases.
  • Selective human DGAT2 inhibitor
  • Inhibits triglyceride (TG) biosynthesis
  • Effectively inhibits lipid droplet formation
  • Useful for research on DGAT2 and TG-related metabolic diseases
Catalog No. 50-004-01384